Would you like to log in again?
FACE SHIELD F60 WIRE 40-LARGE MESH BLACK
Supplier: ORS NASCO, INC.
Face Shield F60 Wire 40-Large Mesh Black^ Jackson Safety* F60 Wire Face Shield, Lens Coating/Shade: Black, Lens Material: Steel Mesh, 40-Large Mesh, Dimensions: 15 1/2 in x 9 in, Sturdy wire face shields, Not intended for high-impact protection, Apply over a face shield to dissipate heat
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F60 WIRE 40-LARGE MESH BLACK 76324-148 138-29081 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty