Would you like to log in again?
FACE SHIELD F30 ACETATE CLR 15 1/2X9 IN
Supplier: ORS NASCO, INC.
Face Shield F30 Acetate Clr 15 1/2X9 In^ Face Shield, Model: 194OU, Lens Material: Acetate, Lens Coating/Shade: clear, Color: Clear, Dimensions: 9 inx15 1/2 in, Thickness: 0.04 in, Unbound, Ideal for scratch resistance and premium optics, High-grade cast sheet acetate protects against splash
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F30 ACETATE CLR 15 1/2X9 IN 76324-152 138-29083 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty