Would you like to log in again?
FACE SHIELD F40 PROPIONATE 915-63 GREEN
Supplier: ORS NASCO, INC.
Face Shield F40 Propionate 915-63 Green^ Face Shield, Lens Coating/Shade: Green-Dark, Lens Material: Propionate, Model: 915-63, Dimension: 9 inx15 1/2 in, Thickness: 0.06 in, Injection molded propionate offers distortion-free optics, 99.9% UV protection, Meets ANSI Z87.1 Standard
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F40 PROPIONATE 915-63 GREEN 76324-156 138-29086 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty