Would you like to log in again?
FACE SHIELD F30 ACETATE 3440 BULK BX50
Supplier: ORS NASCO, INC.
Face Shield F30 Acetate 3440 Bulk BX50^ JACKSON SAFETY* Face Shield, F30 Acetate, Lens Material: Acetate, Color: Clear, Lens Coating/Shade: Clear, Model: 3440 BULK, Dimension: 15 1/2x9 in, Thickness: 0.04 in, Ideal for scratch resistance and premium optics, Meets ANSI Z87.1 Standard
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F30 ACETATE 3440 BULK BX50 76324-164 138-29091 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty