Would you like to log in again?
FACE SHIELD F20 PC CLR 15 1/2 IN 0.04 IN
Supplier: ORS NASCO, INC.
Face Shield F20 Pc Clr 15 1/2 In 0.04 In^ JACKSON SAFETY* Face Shield, F20, Lens Material: Polycarbonate, Color: Clear, Lens Coating/Shade: Clear, Dimension: 8 inx15 1/2in, Thickness: 0.04in, Provide a high degree of protection on the job, Ideal for situations that require impact and heat resistance
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F20 PC CLR 15 1/2 IN 0.04 IN 76324-166 138-29096 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty