Would you like to log in again?
FACE SHIELD ECONOMY F10 PETG 15 1/2X8 IN
Supplier: ORS NASCO, INC.
Face Shield Economy F10 Petg 15 1/2X8 In^ Jackson Safety* Economy Face Shield, Color: Medium Green, Lens Coating/Shade: Medium Green, Series: F10, Material: PETG, Dimensions: 15 1/2x8 in, Thickness: 0.040 in, Lightweight comfort and economical protection, excellent impact strength
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD ECONOMY F10 PETG 15 1/2X8 IN 76324-172 138-29101 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty