Would you like to log in again?
BLANKET WELD-O-GLASS 60 IN X 50 YD RL1
Supplier: ORS NASCO, INC.
Blanket Weld-O-Glass 60 In X 50 Yd RL1^ Weld-O-Glass Blanket, Material: Polyurethane Coated Fiberglass. Dimensions: 60 in X 50 yd, Approved for use in vertical applications with light-moderate exposures resulting from chipping, grinding, heat treating, sand blasting and lilght horizontal welding
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BLANKET WELD-O-GLASS 60 IN X 50 YD RL1 76324-192 138-36149 Roll of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty