Would you like to log in again?
CURTAIN WILSON SEE-THRU WELDING 10X6FT
Supplier: ORS NASCO, INC.
Curtain Wilson See-Thru Welding 10X6Ft^ Wilson See-Thru Welding Curtains, Vinyl, Gold, Heavy-duty vinyl designed to ultraviolet radiation and blue light, Grommeted and ready-to-hang, Thickness: 14 mil, Dimension: 10 ft x 6 ft
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CURTAIN WILSON SEE-THRU WELDING 10X6FT 76324-202 138-36281 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty