Would you like to log in again?
CURTAIN WELDING SPECTRA ORANGE 6 FTX8 FT
Supplier: ORS NASCO, INC.
Curtain Welding Spectra Orange 6 Ftx8 Ft^ See-Thru Welding Curtain, Material: Vinyl, color: Spectra Orange, Dimensions: 6ftX8ft, Heavy-duty vinyl designed to filter far/near ultraviolet radiation and blue light, Grommetted and ready to hang
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CURTAIN WELDING SPECTRA ORANGE 6 FTX8 FT 76324-206 138-36307 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty