Would you like to log in again?
BLANKET WELDING CHIPPING 10 FT X 10 FT
Supplier: ORS NASCO, INC.
Blanket Welding Chipping 10 Ft X 10 Ft^ Welding Blanket, Approved for use in vertical applications with light-moderate exposures resulting from chipping, grinding, heat treating, sand blasting and lilght horizontal welding, Prevents sparks from escaping a confined area, Dimensions: 10 ft X 10 ft
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BLANKET WELDING CHIPPING 10 FT X 10 FT 76324-214 138-37192 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty