Would you like to log in again?
KIT BALDER EXTERNAL SAFETY LENS CLR PK10
Supplier: ORS NASCO, INC.
Kit Balder External Safety Lens PK10^ Balder External Safety Lens Kit, Sturdy polycarbonate external filter plate for use with WH70 BH3 and SmarTIGer helmet. Curved plate offers reduced heat build-up, reflections, and fogging compared to the HSL-100 plate. Lens Tint: Clear, Dimension: 4 1/3x4 1/3in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT BALDER EXTERNAL SAFETY LENS CLR PK10 76324-220 138-37247 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty