Would you like to log in again?
KIT BALDER SMARTIGER SAFETY LENS PK10
Supplier: ORS NASCO, INC.
Kit Balder Smartiger Safety Lens PK10^ Balder SmarTIGer Internal Safety Lens Kit, Prevents surface damage to welding filter and auto-darkening lenses, Color: Clear, Material: Polycarbonate, Used With JACKSON SAFETY* WH70 BH3* Helmet with BALDER* Tech,Dimension: 6 1/2x0.79in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT BALDER SMARTIGER SAFETY LENS PK10 76324-224 138-37249 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty