Would you like to log in again?
FILTER AIRMAX ELITE REPLACEMENT HE CS6
Supplier: ORS NASCO, INC.
Filter Airmax Elite Replacement He CS6^ Airmax Elite Replacement HE Filter, Black, Designed for use only with Jackson Safety Airmax* Elite* PAPR system, Two filters are required for PAPR use, NIOSH approved for particulate contaminants in welding fumes, With 6 HE Filters
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER AIRMAX ELITE REPLACEMENT HE CS6 76324-228 138-40878 Case of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty