Would you like to log in again?
WELDING HELMET ADF 9-13 I2 3.93X2.36IN
Supplier: ORS NASCO, INC.
Helmet Insight Variable Adf Welding 12In^ Insight Digital Variable ADF Welding Helmets, Variable shades to shield and protect eyes from radiant energy during weld, Operates with lithium battery power. Low-battery indicator light, 9-13, Dimension: I2in 3.93x2.36in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WELDING HELMET ADF 9-13 I2 3.93X2.36IN 76324-256 138-46138 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty