Would you like to log in again?
HELMET NEXGEN DIG ADF WELDING ARC ANGEL
Supplier: ORS NASCO, INC.
Helmet Nexgen Dig Adf Welding Arc Angel^ W60 Digital Variable ADF Welding Helmets, Color: Arc Angel, Versatile 3-in-1 function: Weld, Torch, Grind,Ideal for low-amp TIG welding, Material: Polycarbonate/ABS Blend, Used With HSL 100 Shell; no. 16622 - ADF Cartridge, Dimension: 3.8 in x 2.35 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HELMET NEXGEN DIG ADF WELDING ARC ANGEL 76324-268 138-46152 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty