Would you like to log in again?
TRUESIGHT LENS/CARTRIDGES 5 1/4 IN 9-13
Supplier: ORS NASCO, INC.
Truesight Lens/Cartridges 5 1/4 In 9-13^ TrueSight Lens & Cartridges, Variable Shade 9-13, Poly, Amber band pass for enhanced clarity of weld puddle, Built-in magnification lens, Low battery indicator, Color: Black, Material: Polycarbonate, Dimension: 5 1/4 in x 4 1/2 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TRUESIGHT LENS/CARTRIDGES 5 1/4 IN 9-13 76324-276 138-46158 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty