Would you like to log in again?
HELMET TRUESIGHT II DIGITAL HLX BL 4IN
Supplier: ORS NASCO, INC.
Helmet Truesight Ii Digital Hlx Bl 4In^ Truesight II Digital Variable ADF Welding Helmets, Ace of Spades,Battery Type: Alkaline, Material: Thermoplastic, Power Source: AAA Batteries; Solar Cell, Ratchet, Used With HLX100 and HSL100 Shells, Color: Black, Dimension: 3 1/4 in x 4 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HELMET TRUESIGHT II DIGITAL HLX BL 4IN 76324-278 138-46159 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty