Would you like to log in again?
CAP SENTRY III WELDING 6 POINT YL CASE12
Supplier: ORS NASCO, INC.
Cap Sentry Iii Welding 6 Point Yl CASE12^ Sentry III Welding Cap, 6 Point Ratchet, color: Yellow, Dimensions: 6 inx11 1/2 inx8 3/4 in, Material: Polyethylene (HDPE), Size Group: 6 1/2 - 8, Silhouette-style cap, Lightweight construction provides long-term comfort, safety
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP SENTRY III WELDING 6 POINT YL CASE12 76324-284 138-14407 Case of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty