Would you like to log in again?
LINER 325 ULTRA WINTER TWILL/NYLON CS3
Supplier: ORS NASCO, INC.
Liner 325 Ultra Winter Twill/Nylon CS3^ 325 Ultra Winter Liner, Cotton Twill/Nylon Knit, Polyester Fleece Lining, Blue,Ideal for cold-weather environments, Style: Full Neck Flap, Material: Top Shell: Cotton Twill; Bottom Shell: Double Nylon, Meets 16 CFR 1610, Class 1; Meets NFPA 701 Standard
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LINER 325 ULTRA WINTER TWILL/NYLON CS3 76324-296 138-14499 Case of 3 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty