Would you like to log in again?
CONNECTOR QUIK-TRIK CABLE 2/0 AWG FEMALE
Supplier: ORS NASCO, INC.
Connector Quik-Trik Cable 2/0 Awg Female^ Quik-Trik Cable Connector, Connection Type: Single Dome-Nose Connection, Cable Cap.: 1/0-2/0 AWG, Style: Female, camlock and locking bar draw together for virtually unbreakable connection,Form-fitted rubber jacket insulator protects against moisture,oil/dirt
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CONNECTOR QUIK-TRIK CABLE 2/0 AWG FEMALE 76324-310 138-14737 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty