Would you like to log in again?
WRAP-AROUND RULER MEDIUM GRY 3.88INX4FT
Supplier: ORS NASCO, INC.
Wrap-Around Ruler Medium Gry 3.88Inx4Ft^ Wrap-Around Ruler, Medium, Resistance: Heat and Cold Resistant, color: Gray, Dimensons: 3.88 in x 4 ft, Pipe Capacity: 10 in / 3 in, Feature metric scale and pitch chart, Resists heat and cold
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WRAP-AROUND RULER MEDIUM GRY 3.88INX4FT 76324-372 430-14763 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty