Would you like to log in again?
MARKER CONTOUR AL W/WEB STRAP 8IN - 48IN
Supplier: ORS NASCO, INC.
Marker Contour Al W/Web Strap 8In - 48In^ Contour* Marker, Material: Aluminum, Includes: Soapstone Crayon, Web Strap, X-shape frame, calibrated protractor, and a triple joint marking arm and holder, Laterals at any angle, tees, wyes, crosses elbows, Compact, Pipe Capacity: 8 in - 48 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MARKER CONTOUR AL W/WEB STRAP 8IN - 48IN 76324-384 430-14773 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty