Would you like to log in again?
ANGLE LEVEL MAGNETIC 1 VIAL ALUMINUM 9IN
Supplier: ORS NASCO, INC.
Angle Level Magnetic 1 Vial Aluminum 9In^ Contour* Angle Level, Magnetic, Accuracy: 2.5 degrees, Base Type: Grooved, Material: Aluminum, Dimensions: 2 inx9 inx3/4 in, Thickness: 3/4 in, 360-degree adjustable DSL with two 90-degree quadrants graduated in 2-1/2 degree increments, Lightweight
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ANGLE LEVEL MAGNETIC 1 VIAL ALUMINUM 9IN 76324-390 430-14777 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty