Would you like to log in again?
MARKER RADIUS STA ALUMINUM 0.37LB 24 IN
Supplier: ORS NASCO, INC.
Marker Radius Sta Aluminum 0.37Lb 24 In^ Radius Marker, Standard, Radius [Max]: 24 in, Material: Aluminum, Precision tools made of rust proof aluminum and anodized for lasting protection, The arms are precisely aligned with spring tension joints to guarantee accuracy and rigidity
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MARKER RADIUS STA ALUMINUM 0.37LB 24 IN 76324-398 430-14782 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty