Would you like to log in again?
BURNING GUIDE MAGNETIC HEAT/SPATTER N028
Supplier: ORS NASCO, INC.
Burning Guide Magnetic Heat/Spatter N028^ Burning Guides Magnetic, Resistance: Heat and Spatter, Blade Length: 24 in, Size Group: no. 28, Width: 2 in, Use with a burning torch or straight edge, Base has three 30-pound pull cast-in magnets, Withstands heat and spatter, weight: 2.13 lb
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BURNING GUIDE MAGNETIC HEAT/SPATTER N028 76324-406 430-14791 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty