Would you like to log in again?
UNIVERSAL LEVEL 2 INX9.75 INX4.75 IN
Supplier: ORS NASCO, INC.
Universal Level 2 Inx9.75 Inx4.75 In^ Contour* Universal Level, Multi-purpose tool for ensuring accurate leveling, setting predetermined pitch of a string of pipe, setting angle of laterals to precise degree, Accuracy: 2.5 degrees, Size 1: Hole Spaced 2 1/4 - 8 in Centers, Dimensions: 2x9.75x4.75 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
UNIVERSAL LEVEL 2 INX9.75 INX4.75 IN 76324-410 430-14796 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty