Would you like to log in again?
FILTER W40 NITRO AUTO-DARKENING 9-12
Supplier: ORS NASCO, INC.
Filter W40 Nitro Auto-Darkening 9-12^ W40 NITRO Variable Auto-Darkening Filters, Shade 9-12, Features variable shade, sensitivity and delay adjustments, Solar powered, Meets ANSI Z87.1 and CSA Compliant, Three independent sensors, Solar powered
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER W40 NITRO AUTO-DARKENING 9-12 76324-464 138-21947 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty