Would you like to log in again?
KIT NITRO HELMET LENS NITRO ADF 5X2IN
Supplier: ORS NASCO, INC.
Kit Nitro Helmet Lens Nitro Adf 5X2In^ NITRO Helmet Parts, Lens Kits, Allows you to prevent surface damage to welding filter plates and auto-darkening filters, Includes: Nitro 3-Sensor Kit: (2) Internal Safety Plates, (5) Front Safety Plates, Material: Polycarbonate, Dimension: 5 in x 2 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT NITRO HELMET LENS NITRO ADF 5X2IN 76324-466 138-22603 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty