Would you like to log in again?
KIT LENS WELDING SAFETY PLATEW30 ELEMENT
Supplier: ORS NASCO, INC.
Kit Lens Welding Safety Platew30 Element^ Welding Safety Plate W30 ELEMENT* Lens Kit, Clear, Top-quality polycarbonate lenses, Used With Jackson Safety Element ADF no. 23286, Material: Polycarbonate, Color: Clear, with 5 Front, 2 Back, Dimension: 3.5x4.33 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT LENS WELDING SAFETY PLATEW30 ELEMENT 76324-474 138-23440 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty