Would you like to log in again?
CLAMP GROUND 500 A WITH HANDLES
Supplier: ORS NASCO, INC.
Clamp Ground 500 A With Handles^ Ground Clamp, Amps [Nom]: 500 A, Jaw Opening [Nom]: 2 in, Large contact areas, high-clamping pressure and long-levered handles for easy jaw opening, Clamps have fully insulated pivots and springs, The cable jacket is held securely inside conductor body
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLAMP GROUND 500 A WITH HANDLES 76324-562 138-14732 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty