Would you like to log in again?
CAP BC 100 BUMP RATCHET PINLOCK RED HDPE
Supplier: ORS NASCO, INC.
Cap Bc 100 Bump Ratchet Pinlock Red Hdpe^ BC 100 Bump Cap, Adjusting Method: Ratchet, Color: red, Material: HDPE, Suspension: Pinlock, Protects against minor bumps where ANSI standards are not required, Lightweight, Applications: Construction; Light Industrial; Assembly; Manufacturing
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP BC 100 BUMP RATCHET PINLOCK RED HDPE 76324-610 138-14815 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty