Would you like to log in again?
FACE SHIELD F30 ACETATE SCHEDULE B VISOR
Supplier: ORS NASCO, INC.
Shield F30 Acetate Face Clearb Visor 8In^ F30 Acetate Face Shields, Clear, Schedule B Visors, Ideal for scratch resistance and premium optics, High-grade cast sheet acetate protects against splash, 99.9% UV protection, Dimension: 5 1/2 in x 8 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F30 ACETATE SCHEDULE B VISOR 76324-670 138-29054 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty