Would you like to log in again?
FACE SHIELD F60 WIRE MESH STEEL 6 1/2 IN
Supplier: ORS NASCO, INC.
Face Shield F60 Wire Mesh Steel 6 1/2 In^ Jackson Safety* F60 Face Shield, Material: Wire Mesh, Lens Coating/Shade: Mesh Steel, Lens Tint: Mesh Steel, Dimensions: 6 1/2 inx15 1/2 in, Thickness: 0.020 in, Sturdy wire face shields, Not intended for high-impact protection, to dissipate heat
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FACE SHIELD F60 WIRE MESH STEEL 6 1/2 IN 76324-676 138-29102 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty