Would you like to log in again?
RETAINER NEXGEN REPLACEMENT15977 5 1/8IN
Supplier: ORS NASCO, INC.
Retainer Nexgen Replacement15977 5 1/8In^ Nexgen Replacement Retainer, 15977, Retainer lens clip adds another layer of safety between you and the task at hand, Material:Polycarbonate, Used WithJackson HSL-100 with NexGen autodark lens and HLX, Dimension: 6 3/4x5 1/8 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RETAINER NEXGEN REPLACEMENT15977 5 1/8IN 76324-694 138-15977 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty