Would you like to log in again?
FILTER W60 NEXGEN DIG AUTO-DARKENING 1PR
Supplier: ORS NASCO, INC.
Filter W60 Nexgen Digital Auto-Darkening^ W60 NEXGEN Digital Auto-Darkening Filters, Lithium Batteries, Digital controls feature variable shade, sensitivity and delay adjustments, Versatile 3-N-1 built-in function: Weld, Torch, Grind, Built-in mag lens holder, Amber view lens
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER W60 NEXGEN DIG AUTO-DARKENING 1PR 76324-696 138-15984 Pair Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty