Would you like to log in again?
BUMP CAP BC 100 WITH PINLOCK WHITE HDPE
Supplier: ORS NASCO, INC.
Bump Cap Bc 100 With Pinlock White Hdpe^ BC 100 Bump Cap, with Visor Attachment, Pinlock, color: White, Material: HDPE, Dimensions: 5 1/2 inx1 1/2 inx8 1/2 in, Adjusting Method: Ratchet, Applications: Construction; Light Industrial; Assembly; Manufacture, Protects against minor bumps
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BUMP CAP BC 100 WITH PINLOCK WHITE HDPE 76324-704 138-20186 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty