Would you like to log in again?
PLATE INNER SAFETY NITRO HELMET PART CLR
Supplier: ORS NASCO, INC.
Plate Inner Safety Nitro Helmet Part Clr^ Jackson Safety* Inner Safety Plate, NITRO Helmet Part, Lens Coating/Shade: Clear, Lens Tint: Clear, Material: Polycarbonate, Window: 90 x 110 mm, Allows you to prevent surface damage to welding filter plates and auto-darkening filters, Top-quality Pc
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
PLATE INNER SAFETY NITRO HELMET PART CLR 76324-720 138-20755 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty