Would you like to log in again?
BLANKET WELDING SILICA TAN FBRGL 6X8FT
Supplier: ORS NASCO, INC.
Blanket Welding Silica Tan Fbrgl 6X8Ft^ Welding Blankets, Fiberglass, Black, Approved for use in vertical applications with light-moderate exposures resulting from chipping, grinding, heat treating, sand blasting and lilght horizontal welding, Dimension: 6 ft X 8 ft
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BLANKET WELDING SILICA TAN FBRGL 6X8FT 76324-728 138-36318 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty