Would you like to log in again?
FRAME STUR-D-SCREEN STEEL BK 6 FT X 4 FT
Supplier: ORS NASCO, INC.
Frame Stur-D-Screen Steel Bk 6 Ft X 4 Ft^ Stur-D-Screen Frame, Coating: Powder coated, Material: Steel, color: black, No. Of Panels: 1, Dimensions: 6 ft X 4 ft, multiple configurations for a variety of work environments, Includes: Square Steel Frame, Lacing Ties, Platform T Legs, Brackets
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FRAME STUR-D-SCREEN STEEL BK 6 FT X 4 FT 76324-730 138-36335 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty