Would you like to log in again?
KIT 384-WL BIOPRINTING BK UCLEAR PLT CS1
Supplier: Greiner Bio-One
Kit 384-Wl Bioprinting Bk uclear Plt CS1^ Kit 384 Well BiO Assay, Includes: 600 ul vials of NanoShuttle*-PL, 6Well Levitating/384well Spheroid/Holding/Ring Drive, 384Well Deep Well Plate, 6Well Culture Multiwl Plates with cell-repellent surface, 384Wl Culture Microplate with cell-repellent surface
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT 384-WL BIOPRINTING BK UCLEAR PLT CS1 76324-770 781841 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty