Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HyClone™ Standard Fetal Bovine Serum (FBS), U.S. Origin, Cytiva
Supplier: HyClone products (Cytiva)
Hyclone™
SH30088.02HI SH30088.02 SH30088.03 SH3008803IH25-40 SH30088.02HI SH30088.03HI SH30088.03HI SH30088.03IR25-40 SH30088.02 SH30088.03 SH30088.03IR25-40 SH3008803IH25-40
82013-574EA 833.69 USD InStock
82013-574 76236-308 76324-888 CA76324-888 CA76236-308 CA16777-534 CA76324-890 CA16777-532 16777-534 76324-890 CA82013-574 16777-532
HyClone™ Standard Fetal Bovine Serum (FBS), U.S. Origin, Cytiva
Sera (Serum)
HyClone™ standard cell culture grade FBS is filtered through three sequential 100 µm (0,1 µm) pore-size rated filters.
  • Sourced in the USA with complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters
HyClone Standard Fetal Bovine Serum (FBS), U.S. origin is an economical alternative to characterized and defined FBS. Standard FBS is ideal for research applications.
Standard FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC) and IgG. Included assays are subject to change without notice.
Certifications: Virus panel testing according to 9 CFR 113.53.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty