Would you like to log in again?
HyClone® Cosmic Calf Serum, U.S. Origin
Supplier: HyClone products (Cytiva)
HyClone™
SH30087.03IR25-40 SH30087.04HI SH30087.03IR25-40 SH30087.02 SH30087.03HI SH30087.02HI SH30087.04HI SH30087.04IR30-45 SH30087.02 SH30087.04IR25-40 SH30087.04IR SH30087.04 SH30087.04IR25-40 SH30087.02HI SH30087.03 SH30087.03HI SH30087.03 SH30087.04 SH30087.04IR30-45
82007-442EA 250.97 USD InStock
82007-442 82013-582 CA82007-442 76324-884 CA76324-884 76324-930 76324-886 CA16777-242 16777-244 16777-242 CA76324-886 CA76324-930 CA76234-774 76234-776 CA76234-776 82013-614 CA82013-582 76234-774 CA16777-244
HyClone® Cosmic Calf Serum, U.S. Origin
Sera (Serum)
HyClone™ Cosmic calf serum is fortified with iron and naturally derived components for increased performance with a broad spectrum of cell type. Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.
  • Filtered through sequential 100 nm pore size rated filters
  • Extensive quality, performance and analytical testing
All components are sourced in the U.S. and are traceable. Studies have shown Cosmic Calf outperforms iron-supplemented bovine calf serum and FBS in several applications, and performs exceptionally well with CHO-KI cells and CHO derivatives.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty