Would you like to log in again?
Primate Enrichment Treats, Acacia Gum, NF/FCC
Supplier: Bio-Serv
F7015 F7015
76325-044BG 63.83 USD InStock
76325-044 CA76325-044
Primate Enrichment Treats, Acacia Gum, NF/FCC
Animal Supplies Animal Environmental Enrichment
Made from the sap of acacia trees. Acacia gum should be a moist and sticky consistency, but not runny, for easier application.
  • Natural form of edible enrichment
  • Add foraging treats and bits for added interest
  • Place in toys and wood sticks to encourage foraging behavior
Shelf life: 12 months, cool dry storage.
Bio-Serv's flavored PRANGTM can be substituted for the water to provide added variety.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty