Would you like to log in again?
HiTrap Column, Capto Core 400
Supplier: Cytiva
HiTrap™
17372411 17-3724-11
76325-056EA 503.81 USD InStock
76325-056 CA76333-722
HiTrap Column, Capto Core 400
Chromatography Columns
Novel core bead technology and multimodal ligand give Capto Core 400 dual functionality, size exclusion and binding chromatography, in one chromatography resin.
  • High capacity and productivity in flowthrough mode
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification
  • Wide operational window of pH and conductivity
Capto Core 400 is an excellent alternative to size‑exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty