Would you like to log in again?
HiScreen™ Capto Core 400, Multimodal Columns, Cytiva
Supplier: Cytiva
HiScreen™
17-3724-10
76325-058EA 298.48 USD InStock
76325-058
HiScreen™ Capto Core 400, Multimodal Columns, Cytiva
Chromatography Columns
HiScreen™ columns prepacked with Capto Core 400 are designed for intermediate purification and polishing of viruses and other large biomolecules. Novel core bead technology and multimodal ligand give Capto Core 400 dual functionality, size exclusion and binding chromatography, in one chromatography resin.
  • For intermediate purification and polishing of viruses and other large biomolecules (Mr >350000) in flowthrough mode
  • Novel core bead technology allows efficient capture of contaminants, while target molecule is collected in the flowthrough
  • Significantly improved productivity and higher flow rates compared with gel filtration methods
  • Straightforward optimisation due to flowthrough chromatography and robust performance
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap™ and HiScreen™ columns
Capto Core 400 is an excellent alternative to size‑exclusion chromatography (SEC) resins that are typically employed in the final stages of virus purification in vaccine manufacture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty