Would you like to log in again?
Thermo Sequenase DNA Polymerase, Cytiva
Supplier: Cytiva
E79000Z E79000Y E79000Z E79000Y
76325-070EA 1231.22 USD InStock
76325-070 CA76325-072 76325-072 CA76325-070
Thermo Sequenase DNA Polymerase, Cytiva
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Thermo Sequenase is a novel thermostable DNA polymerase that uses dideoxynucleotide triphosphates (ddNTPs) as readily as deoxynucleotide (dNTP) substrates. This results in extremely uniform and easy-to-read sequence band patterns. It is an excellent choice for cycle sequencing where template quantity limitations preclude the use of T7 Sequenase DNA polymerase.
  • One unit of enzyme incorporates 10 nmol of dNTPs into acid-insoluble material in 30 min at 74°C
  • For DNA Sequencing and SNP genotyping applications where primer extension is employed
  • Uniform and easy-to-read band patterns
  • Thermostable enzyme
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty