Would you like to log in again?
Actero™ EZ-Media Bag, FoodChek
Supplier: FoodChek Systems
Actero™ Actero™ EZ
FCM-259 FCM-263 FCM-256 FCM-258 FCM-261 FCM-257
76325-092EA 2879.25 USD InStock
76325-092 76325-096 76325-094 76325-100 76325-098 76325-102
Actero™ EZ-Media Bag, FoodChek
Bags Media Bags
The Actero™ EZ-Media Bag contains high quality, pre-measured and sterilized enrichment media powder that greatly reduces the typical steps and glassware required in traditional media preparation.
  • Increases safety – Contains sterile enrichment medium that eliminates autoclaving or hot glassware
  • Eliminates human error – Contains pre-weighed medium, just add deionized water through the filter (ordered separately) and gently massage the bag to mix
  • Reduces time – Eliminates measuring, heating or standard consumables
  • Improves testing throughput – Quick to rehydrate the powder medium through the filter (ordered separately) and quick to dispense
  • Lower cost – Compact powder format and packaged in a flat bag to reduce time to prepare, shipping costs and storage requirements
Actero™ EZ-Media Bags are available in the enrichment mediums of:
Demi-Fraser - Demi (Half) Fraser is a modification of Fraser Broth base. This media was developed by Fraser and Sperber for the rapid detection of Listeria from food and environmental samples. The addition of ferric ammonium citrate allows for the selective enrichment of Listeria spp.
The patented Actero™ Elite Listeria Enrichment Media - A selective medium specifically optimized for single-step recovery and enrichment of Listeria spp. from food and environmental samples.
Buffered Peptone Water ISO - Used for pre-enriching damaged Salmonella spp. from various food sources.
Ordering information: To use, just add deionized water and gently massage the bag to rehydrate. There are 15 dry media bags per Each and includes two 0.2 µm filters. Each filter can be used up to 10 times.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty