Would you like to log in again?
LABEL CRYO 0.75IN X 4IN INKJET PRINT RL1
Supplier: GA INTERNATIONAL INC
Label Cryo 0.75In X 4In Inkjet Print Rl1^ Cryo-JetTAG* -Cryogenic Labels for Inkjet Printers; Color: White, Labels/roll: 600, Core: 1in, Perforation: No, Corners: Round; Corners: round, Temperature range: from -196ºC to +110ºC. Size: 0.75in x 4in 19.1mm x 101.6mm Labels across: 2
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LABEL CRYO 0.75IN X 4IN INKJET PRINT RL1 76325-916 807656032344 Roll of 1 Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty