Would you like to log in again?
NEBNext® Cell Lysis Buffer Module, New England Biolabs
Supplier: New England Biolabs
NEBNext®
E5530S
76326-030EA 106 USD InStock
76326-030
NEBNext® Cell Lysis Buffer Module, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
This module contains the enzymes and buffers required to lyse isolated cultured and primary cells to extract RNA. The module is optimized for use with the NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina or NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module, and can be used to supplement the NEBNext Cell Lysis Buffer and Murine RNase Inhibitor components supplied in these products.
  • Includes NEBNext Cell Lysis Buffer
  • Includes Murine RNase Inhibitor
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty