Would you like to log in again?
Nuclease P1
Supplier: New England Biolabs
M0660S
76326-036EA 57 USD InStock
76326-036
Nuclease P1
Enzymes
Nuclease P1 (from P. citrinum) is a zinc-dependent single-strand specific nuclease which hydrolyzes 3' to 5' phosphodiester bonds in RNA and ssDNA with no base specificity.
  • Single-stranded (ssDNA or RNA) specific endonuclease
  • Conversion of single-stranded DNA or RNA to 5'-mononucleotides
  • Analysis of the base composition of nucleic acids
  • Studies of the potential damage and modification in DNA
Nuclease P1 also exhibits 3-phosphomonoesterase activity. Although a single-strand specific nuclease (ssDNA and RNA-specific), Nuclease P1 does display some activity toward dsDNA in Nuclease P1 Reaction Buffer. I preferentially degrading single-stranded nucleic acids (ssDNA or RNA) in the presence of double stranded DNA (dsDNA), we recommend using Nuclease P1 in 1X NEBuffer 1.1 to limit activity on dsDNA while maintaining single-strand nuclease activity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty